Learn More
Tocris Bioscience™ Kobe 0065 Available on GSA/VA Contract for Federal Government customers only.
H-Ras-cRaf1 interaction inhibitor; inhibits Raf signaling
$139.00 - $585.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
54-751-0 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5475/10
Each for $139.00
Add to cart
54-755-0 View Documents Tocris Bioscience™
5475/50
Each for $585.00
Add to cart
Description
Description
H-Ras-cRaf1 interaction inhibitor; inhibits Raf signaling. Selectively attenuates proliferation and induces apoptosis of cancer cells harboring Ras oncogenes. Reduces growth of K-Ras mutant SW480 colon carcinoma cell xenografts in mice. Orally active.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.