Carestream Health BioMax™ Autoradiography Cassettes
Maximize image resolution and background clarity by ensuring intimate contact between the film and sample in a light-tight environment
Manufacturer: Carestream Health 835 6800
 View more versions of this product
Catalog No. 05-728-61
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    For Use With (Application) Maximize image resolution and background clarity by ensuring intimate contact between the film and sample in a light-tight environment
    Includes Cassette with BioMax TranScreen HE Intensifying Screen
    Product Type Autoradiography Cassette
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback