Learn More
Carestream Health BioMax™ Autoradiography Cassettes
Maximize image resolution and background clarity by ensuring intimate contact between the film and sample in a light-tight environment
Manufacturer:  Carestream Health 857 8825
 View more versions of this product
Catalog No. 05-728-16
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Autoradiography Cassette
Maximize image resolution and background clarity by ensuring intimate contact between the film and sample in a light-tight environment
Cassette with BioMax MS Intensifying Screen
Accessories