Carestream Health BioMax™ Autoradiography Cassettes
Maximize image resolution and background clarity by ensuring intimate contact between the film and sample in a light-tight environment
Manufacturer: Carestream Health 875 0499
 View more versions of this product
Catalog No. 05-728-17
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Product Type Autoradiography Cassette
    For Use With (Application) Maximize image resolution and background clarity by ensuring intimate contact between the film and sample in a light-tight environment
    Includes Cassette with BioMax MS Intensifying Screen
    • Available with or without BioMax or TranScreen™ intensifying screen BioMax MS and TranScreen HE intensifying screens offer maximum sensitivity for 32P and 125I
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback