Learn More
Carestream Health BioMax™ MR (Maximum Resolution) Film
Recommended for direct autoradiography of weak and medium energy β-radioisotopes, such as 35S,33P, and 14C
Manufacturer:  Carestream Health 894 1114
 View more versions of this product
Catalog No. 05-728-26
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
5 x 7 in. (12.7 x 18cm)
12.7cm
5 in.
7 in.
Provides maximum resolution and sensitivity for detection of 35 S, 33 P, and 14 C chemiluminescent labeled samples, ideal for the detection of sequencing gels coated on only one side of a clear base Utilizes patented “T-grain” technology
18cm
BioMax MR-1 Film: Noninterleaved
50 Pack; Bulk Pack