Learn More
Carestream Health BioMax™ MR (Maximum Resolution) Film
Recommended for direct autoradiography of weak and medium energy β-radioisotopes, such as 35S,33P, and 14C
Manufacturer:  Carestream Health 8952855
 View more versions of this product
Catalog No. 05-728-31
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
8 x 10 in. (20.32 x 25cm)
20.32cm
8 in.
10 in.
Provides maximum resolution and sensitivity for detection of 35 S, 33 P, and 14 C chemiluminescent labeled samples, ideal for the detection of sequencing gels coated on only one side of a clear base Utilizes patented “T-grain” technology
25cm
BioMax MR-2 Film: ReadyPacked
50 Pack; Individually wrapped