Learn More
Carestream Health BioMax™ TranScreen™ HE Autoradiography Intensifying Screens
Innovative intensifying screen developed specifically for detection of higher energy radioisotopes such as 32P and 125I
$372.53
Specifications
For Use With (Application) Detection of higher energy radioisotopes such as 32P and 125I
For Use With (Equipment) BioMax MS film
Product Type Intensifying Screen
View More Specs
Products 1
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
05-728-63 Non-distribution product as customer accommodation.
View Documents
Carestream Health
856 3959
8 x 10 in. (203 x 254 mm)
Each for $372.53
Case of 5 EA for $1,625.86
Add to cart
Description
Description
Equals the sensitivity of the BioMax™ MS screen with improved handling features and publication-quality images
Order Info
Available in two formats: Intensifying screen only; or with BioMax cassette (see 05-728-61).
Specifications
Specifications
Detection of higher energy radioisotopes such as 32P and 125I
BioMax MS film
Intensifying Screen