Learn More
Carestream Health BioMax™ TranScreen™ LE Autoradiography Intensifying Screens
Intensifying screen designed for 3H, 14C, 33P, 35S, 45Ca, and 59Fe
Manufacturer:  Carestream Health 162 2034
Catalog No. 05-728-67
Add to cart
Description
Description
  • Achieve a 0.301 O.D. image five or more times faster than direct autoradiography
  • Offer greater resolution (2X) in the same time with equal or better sensitivity as BioMax XAR/calcium tungstate intensifying screen systems, when used with 32P and 125I
Order Info
Available in two formats: Intensifying screen only; or with BioMax cassette (see 05-728-65).
Specifications
Specifications
Intensifying Screen
203 mm
254 mm
32P and 125 I
8 in.
10 in.
8 x 10 in. (203 x 254 mm)
BioMax MS film