Learn More
Carestream Health BioMax™ XAR™ Film
Suitable for use with all radioisotopes and chemiluminescent probes
Manufacturer:  Carestream Health 165 1579
 View more versions of this product
Catalog No. 05-728-41
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
8 in.
10 in.
For all radioisotopes and chemiluminescent probes, ideal for both direct autoradiography and exposures with intensifying screens utilizes an improved formulation
25cm
X-OMAT AR-2 Film
8 x 10 in. (20 x 25cm)
20cm
50 Pack- Individually wrapped