Learn More
Carestream Health BioMax™ XAR™ Film
Suitable for use with all radioisotopes and chemiluminescent probes
Manufacturer:  Carestream Health 166 0760
 View more versions of this product
Catalog No. 05-728-43
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
5 x 7 in. (13 x 18cm)
13cm
5 in.
7 in.
For all radioisotopes and chemiluminescent probes, ideal for both direct autoradiography and exposures with intensifying screens utilizes an improved formulation
18cm
X-OMAT AR-2 Film
50 Pack- Individually wrapped