Learn More
Carestream Health BioMax™ XAR™ Film
Suitable for use with all radioisotopes and chemiluminescent probes
$395.89 - $1723.28
Specifications
For Use With (Application) For all radioisotopes and chemiluminescent probes, ideal for both direct autoradiography and exposures with intensifying screens utilizes an improved formulation
View More Specs
Products 5
Catalog Number Mfr. No. Dimensions (LxW) Product Type Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (LxW) Product Type Quantity Price Quantity & Availability  
05-728-34 Non-distribution product as customer accommodation.
View Documents
Carestream Health
165 1454
8 x 10 in. (20 x 25 cm) X-OMAT AR-5 Film 50 Pack, Bulk Pack
Pack of 50 for $579.30
Case of 5 PK for $2,571.91
Add to cart
05-728-35 Non-distribution product as customer accommodation.
View Documents
Carestream Health
165 1512
14 x 17 in. (35 x 43 cm) X-OMAT AR-5 Film 50 Pack, Bulk Pack
Pack of 50 for $1,460.05
Case of 5 PK for $6,759.50
Add to cart
05-728-36 Non-distribution product as customer accommodation.
View Documents
Carestream Health
165 1496
5 x 7 in. (13 x 18 cm) X-OMAT AR-5 Film 50 Pack, Bulk Pack
Pack of 50 for $395.89
Case of 5 PK for $1,727.83
Add to cart
05-728-42 Non-distribution product as customer accommodation.
View Documents
Carestream Health
165 1678
14 x 17 in. (35 x 43 cm) X-OMAT AR-2 Film 50 Pack, Individually wrapped
Pack of 50 for $1,723.28
Case of 2 PK for $3,346.18
Add to cart
05-728-43 Non-distribution product as customer accommodation.
View Documents
Carestream Health
166 0760
5 x 7 in. (13 x 18 cm) X-OMAT AR-2 Film 50 Pack, Individually wrapped
Pack of 50 for $678.84
Case of 5 PK for $3,013.83
Add to cart
Description
Description
  • Coated on both sides of a clear base, making it ideal for both direct autoradiography and exposures with intensifying screens Utilizes an improved formulation that eliminates the need for interleaving paper while maintaining the same performance as the original X-Omat™ AR (XAR) film Suitable for manual and automated processing
Order Info
Formats include: Bulk-packed film; ReadyPacked film (with each sheet of film individually wrapped in a protective folder for easy handling)
Specifications
Specifications
For all radioisotopes and chemiluminescent probes, ideal for both direct autoradiography and exposures with intensifying screens utilizes an improved formulation