Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Koehler™ Bright Star™ Submersible Responder™ Flashlights

UL listed for use in hazardous locations, these watertight flashlights are backed by a limited lifetime warranty

Manufacturer:  Koehler™ Bright Star™ 200201

 View more versions of this product

Catalog No. 19-337-640


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

5.66 hr.
30000
Push-button
5.8cm
15 hr.
17cm
Xenon
Orange
6.7 oz.
C Alkaline
UL
2.31 in.
For use in hazardous locations, these watertight flashlights are backed by a limited lifetime warranty
6.75 in.
Polycarbonate
Nylon
Flashlight
0.18kg
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit