Koehler™ Bright Star™ Submersible Responder™ Flashlights

UL listed for use in hazardous locations, these watertight flashlights are backed by a limited lifetime warranty

Manufacturer: Koehler™ Bright Star™ 200203

 View more versions of this product

Catalog No. 19-337-642

  Check Availability
Add to Cart

    Specifications

    Battery C Alkaline
    Certifications/Compliance UL
    Controller Type Push-button
    Diameter (English) Outer 2.31 in.
    For Use With (Application) For use in hazardous locations, these watertight flashlights are backed by a limited lifetime warranty
    Length (English) 6.75 in.
    Length (Metric) 17cm
    Light Source Xenon
    Material Nylon
    Color Black
    Weight (English) 6.7 oz.
    Weight (Metric) 0.18kg
    Battery Run Time 5.66 hr.
    Candle Power 30000
    Diameter (Metric) Outer 5.8cm
    Lamp Life 15 hr.
    Lens Material Polycarbonate
    Product Type Flashlight

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit