Learn More
Koehler™ Instrument Cloud and Pour Point Chamber
Supplier: Koehler™ Instrument K46001
Specifications
Cloud and Pour Point Chamber | |
Inlet and outlet connections, removable composition top plate, 0.5 in. cork insulation around interior, copper jackets, gaskets, disks and thermometer holders | |
ASTM D97, D2500, D5853, D6074, D6158, IP 15, 219, ISO 3015, 3016, DIN 51597, FTM 791-201, NF T 60-105 |
Steel, Copper | |
Polyurethane enamel | |
Circulating refrigerated coolant from external source |