Learn More
Koehler™ Instrument Existent Gum Evaporation Bath
Tests for gum content formed during fuel storage and deposited on induction system surfaces, intake valves, stems and guides.
Supplier: Koehler™ Instrument K33781
Description
- Electrically heated baths determine existent gum in aircraft turbine fuels by steam-jet evaporation and in motor and aviation gasolines by air-jet evaporation
- Fully insulated, aluminum block design assures safe, efficient high temperature operation
- Equipped with air/steam pressure regulator; stainless steel jets deliver air or steam flow to test wells
- Microprocessor PID control provides quick temperature stabilization without overshoot
- Overtemperature control circuit interrupts power should bath temperature exceed a programmed cut-off point
- Dual LED displays provide actual and setpoint temperature values in °C/°F format
Compliance
Complies with ASTM D381; IP 131; ISO 6246; DIN 51784; FTM 791-3302; and NF M 07-004 specifications.
Specifications
Superheater (0-1500w) | |
220-240V 50/60Hz, Single Phase, 6.8A | |
6.8 A |
3 Unit | |
220/240 V | |
50/60 Hz |