Learn More
Koehler™ Four-Unit Reid Vapor Pressure Baths
Manufacturer:  KOEHLER INSTRUMENT COMPANY INC K11450
 View more versions of this product
Catalog No. 13-626-150
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
115V
91.5cm
47.7kg
18.8A
ASTM D323, D1267; GPA 2140; IP 69, 161; ISO 3007, 4256; DIN 51616, 51754; FTM 791-1201; NF M 07-007, 41-010
67 lb.
±0.1°C
36 in.
105 lb.
15 in.
51.9L
100°C
30.4kg