Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Koehler™ Instrument Group 1-3 Front View Distillation Apparatus, Left-Hand Model

For distillation of gasolines, fuels, and other petroleum products

Manufacturer:  Koehler™ Instrument K45100

Catalog No. 11-302-30A


Add to cart

Description

Description

The sample is evaporated and condensed under controlled conditions, and observations are made of the temperatures at which various percentages are recovered and/or the percentages recovered at specified temperatures.

  • Includes Types A and C Flask Support Boards and graduate support block
  • Also available in Right-Hand model, and Group 4 Distillation
Specifications:
  • Electrical Reqts: 115V 50/60Hz
  • Shipping Weight: 65 lb. (29.5kg)
  • Shipping Dimensions: 13.3 cu. ft.

Specifications

Specifications

ASTM D86, E133
19.5 in.
Types A and C flask support bards and graduate support block
460mm
Distillation of gasoline, fuels, and other petroleum products
500mm
18.25 in.
Group 1-3 Front View Distillation Apparatus
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit