Koehler™ Instrument Copper Strip Tarnish Test Accessories

Various accessories associated with Copper Strip Tarnish Test

Manufacturer: KOEHLER INSTRUMENT COMPANY INC 332004001

 View more versions of this product

Catalog No. 14-210-45

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) Designed for use performing astm d4048 cloud pour testing
    For Use With (Equipment) Koehler™ K25330 and K25339 copper test strip baths or Koehler™ K25308 test jar rack
    Color Clear
    Product Type Test Jar

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit