Koehler™ KV5000 Kinematic Viscosity Bath

Provides automatic viscosity measurements of petroleum and petrochemical products

Manufacturer: KOEHLER INSTRUMENT COMPANY INC K23798OS

 View more versions of this product

Catalog No. 13-626-107

  Check Availability

This item is a non-standard voltage; please ensure you are ordering the correct product.

Please contact customer service for assistance: 1-800-766-7000.

Add to Cart

    Specifications

    Hertz 50/60Hz
    Model KV5000
    Temperature Range (Metric) Ambient to 150°C; sub-ambient to 10°C with external cooling
    Voltage 220-240V
    Depth (Metric) 45.7cm
    Depth (English) 18 in.
    Electrical Requirements 220-240V 50/60Hz

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit