Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Koehler™ Instrument KV5000 Kinematic Viscosity Bath

Provides automatic viscosity measurements of petroleum and petrochemical products

$17020.00 - $18600.00

Specifications

Model KV5000
View More Specs

Products 4
Catalog Number Mfr. No. Depth (English) Depth (Metric) Electrical Requirements Price Quantity & Availability  
Catalog Number Mfr. No. Depth (English) Depth (Metric) Electrical Requirements Price Quantity & Availability  
13-626-106
View Documents
Koehler™ Instrument
K23708OS
18 in. 45.7 cm 220/240 V 50/60 Hz
Each for $18,000.00
Add to cart
 
13-626-104
View Documents
Koehler™ Instrument
K23702OS
12 in. 30.4 cm 115 V 60 Hz
Each for $17,020.00
Add to cart
 
13-626-105
View Documents
Koehler™ Instrument
K23792OS
12 in. 30.4 cm 220/240 V 50/60 Hz
Each for $18,270.00
Add to cart
 
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
13-626-107
View Documents
Koehler™ Instrument
K23798OS
18 in. 45.7 cm 220/240 V 50/60 Hz
Each for $18,600.00
Add to cart
 
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Description

Description

Kinematic viscosity is of primary importance in the design and selection of a wide range of petroleum products. Calibrated capillary viscometers are used to measure flow under gravity or vacuum at precisely controlled temperatures.

  • With Optical Flow Detection System
  • Includes communication and power ports for each optical detection assembly; can utilize up to five optical assemblies (K23780 series)
  • Two additional positions are available for manual viscosity measurements; all positions can be used in manual mode
  • Modular design of three types of optical assemblies allows interchangeable use of any combination and size of Cannon™-Fenske, Reverse Flow, and Ubbelohde tubes in viscosity baths
  • Integrated automatic timing on LCD microcomputer with optical sensors automatically measures sample efflux times
  • Automatic calculation and display of results in viscosity units or seconds
  • Bath Medium: water or suitable heat transfer fluid
  • Thermometers sold separately

Compliance

Complies with ASTM D445, D2170, D6074, D6158; IP 71, 319; ISO 3104; DIN 51550; FTM 791-305; and NF T 60-100 specifications.

Specifications

Specifications

KV5000
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit