Koehler Viscosity Reference Standards

For calibration and verification of kinematic and dynamic viscosity test equipment, both manual and automatic

Manufacturer: KOEHLER INSTRUMENT COMPANY INC 355003005

 View more versions of this product

Catalog No. 13-617-54

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Certifications/Compliance ISO/IEC 17025, ASTM D2162, NIST
    For Use With (Equipment) Kinematic and dynamic viscosity test equipment, both manual and automatic
    Product Type Viscosity Standard
    Format Liquid
    For Use With (Application) For calibration and verification of kinematic and dynamic viscosity test
    Material High stability base oils and polybutenes
    Precision ±0.2%
    Viscosity Rating D5
    Viscosity 4.2mm2/sec. at 40°C
    Volume (English) 17 oz.
    Volume (Metric) 500mL

    · Standards are based upon the NIST value of 1.0034 cSt for water at 20°C (68°F)
    · Traceable to National Standards; manufactured according to ASTM D2162 specifications
    · Supplied with ISO/IEC 17025 Certification Report
    · Fully compliant with ASTM and related test procedures
    · Calibrated to a precision of ±0.2% for viscosity and kinematic viscosity
    · Each standard is calibrated at a minimum of five temperatures
    · Supplied in 500mL amber bottle, with full certification and MSDS

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit