Koehler Viscosity Reference Standards

For calibration and verification of kinematic and dynamic viscosity test equipment, both manual and automatic

Manufacturer: KOEHLER INSTRUMENT COMPANY INC 355003103

 View more versions of this product

Catalog No. 13-617-65

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Format Liquid
    Material High stability base oils and polybutenes
    Volume (Metric) 500mL
    For Use With (Equipment) Kinematic and dynamic viscosity test equipment, both manual and automatic
    Certifications/Compliance ISO/IEC 17025, ASTM D2162, NIST
    For Use With (Application) For calibration and verification of kinematic and dynamic viscosity test
    Precision ±0.2%
    Viscosity Rating D1000
    Viscosity 418mm2/sec. at 40°C
    Volume (English) 17 oz.
    Product Type Viscosity Standard

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit