Learn More
Koehler™ Wickbold Combustion Apparatus
Manufacturer:  KOEHLER INSTRUMENT COMPANY INC K47590
 View more versions of this product
Catalog No. 13-626-156
  Check Availability
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Add to cart
Specifications Description & Specifications
Specifications
220 to 240V, 50/60Hz
Total Sulfur in liquefied petroleum (LP) gases
47cm
18.1kg
40 lb.
Determines total sulfur in liquefied petroleum (LP) gases and in liquid petroleum products by the Wickbold Oxy-hydrogen burner method
18.5 in.
220 to 240V
13 in.
ASTM D2384, D2747, D2784, D2785; GPA 2140; IP 243; ISO 4260; DIN EN 41; NF T 60-142