Learn More
Koehler™ Instrument Wickbold Combustion Apparatus
$26104.00
Specifications
Analytical Method Total Sulfur in liquefied petroleum (LP) gases
Certifications/Compliance ASTM D2384, D2747, D2784, D2785; GPA 2140; IP 243; ISO 4260; DIN EN 41; NF T 60-142
For Use With (Application) Determines total sulfur in liquefied petroleum (LP) gases and in liquid petroleum products by the Wickbold Oxy-hydrogen burner method
Height (English) 18.5 in.
Height (Metric) 47 cm
View More Specs
Products 2
Catalog Number Mfr. No. Electrical Requirements Voltage Price Quantity & Availability  
Catalog Number Mfr. No. Electrical Requirements Voltage Price Quantity & Availability  
13-626-155
View Documents
Koehler™ Instrument
K47500
115 V 50/60 Hz 115 V
Each for $26,104.00
Add to cart
13-626-156
View Documents
Koehler™ Instrument
K47590
220/240 V 50/60 Hz 220/240 V This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
Specifications
Specifications
Total Sulfur in liquefied petroleum (LP) gases
Determines total sulfur in liquefied petroleum (LP) gases and in liquid petroleum products by the Wickbold Oxy-hydrogen burner method
47 cm
18.1 kg
ASTM D2384, D2747, D2784, D2785; GPA 2140; IP 243; ISO 4260; DIN EN 41; NF T 60-142
18.5 in.
40 lb.
13 in.