Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kojic acid, 99%, ACROS Organics™
Click to view available options
Quantity:
5g
25g
100g
500g
Packaging:
Glass bottle
Specifications
Quantity | 100g |
Melting Point | 151.0°C to 156.0°C |
Color | Undesignated |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Assay Percent Range | 99% |
Beilstein | 18,II,57 |
Merck Index | 15,5365 |
Solubility Information | Solubility in water: soluble. Other solubilities: freely soluble in acetone and ethanol;,sparingly soluble in diethyl ether,ethyl acetate,,chloroform and pyridine;,insoluble in benezne |
Formula Weight | 142.11 |
Show More |
Safety and Handling
GHS H Statement
Suspected of causing cancer.
GHS P Statement
Obtain special instructions before use.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.
GHS Signal Word: Warning
EINECSNumber : 207-922-4