Learn More
DWK Life Sciences Kimble Kontes Adapter Bushing
Allow for compact assemblies of dissimiliar joints
$53.75 - $153.00
Specifications
Certifications/Compliance | USP Type I, ASTM E438, Type I, Class A |
---|---|
Product Type | Bushing Adapter |
For Use With (Application) | For compact assemblies of dissimilar joints |
Material | Glass |
Catalog Number | Mfr. No. | Standard Taper Joint No. | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Standard Taper Joint No. | Price | Quantity & Availability | |||||
K150750-0265
|
DWK Life Sciences
1507500265 |
24/40 Small, 29/42 Large |
Case for $67.75
|
|
K150750-0242
|
DWK Life Sciences
1507500242 |
14/35, 14/20 Small, 24/40 Large |
Case for $67.75
|
|
K150750-0266
![]() |
DWK Life Sciences
1507500266 |
24/40 Small, 34/45 Large |
Case for $89.00
|
|
K150750-0268
|
DWK Life Sciences
1507500268 |
24/40 Small, 45/50 Large |
Case for $153.00
|
|
K273500-0022
|
DWK Life Sciences
2735000022 |
14/20 Small, 19/22 Large |
Case for $53.75
|
|
Description
- Bushing type enlarging adapter features inner and outer joints for a compact way of changing joint sizes
- Use to connect dissimilar standard taper joints
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
USP Type I, ASTM E438, Type I, Class A | |
Bushing Adapter | |
For compact assemblies of dissimilar joints | |
Glass |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.