Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Please call 1-800-766-7000 to place your order or try our site again later.
DWK Life Sciences Kimble Kontes Butyl Rubber Stopper, Plug style
$231.00
Specifications
Color | Gray |
---|---|
Material | Rubber |
For Use With (Application) | Low temperature applications |
For Use With (Equipment) | Fits GPI aluminum seal finish vials |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
03-340-71Z
|
DWK Life Sciences
7382713 |
13 |
Case of 1000 for $231.00
|
N/A |
Specifications
Gray | |
Rubber | |
Low temperature applications | |
Fits GPI aluminum seal finish vials |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title