Learn More
DWK Life Sciences Kimble™ Kontes™ 14/20 Distillation Cow-Type with 3 Receivers
Cow-type design having three receivers spaced 45° apart.
Supplier: DWK Life Sciences 2878000000
Description
- Distillation receivers 14/20 cow type with three receivers.
- Cow-type design having three receivers spaced 45° apart
- Condensers with drip joints will protrude into the spherical section, keeping wetted surfaces and product hold-up to a minimum
- Fits tubing ID 1/4 IN
- Standard Taper joints 14/20, Standard Taper joint to condenser 14/20
- Case of 1
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Three receivers, No. 1 hose connection | |
Cow Type Distillation Receiver | |
USP Type I, ASTM E438, Type I, Class A | |
14/20 |