Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kimble™ Kontes™ Heavy Wall Recovery Flasks with Square Bead, standard taper Outer Joint

Ideal for use with rotary evaporators

Manufacturer:  Kimble™ KC25165500

 View more versions of this product

Catalog No. 12-141-344


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Yes
95mm
Rotary evaporator
17cm
Single
500mL
3.74 in.
Plastic coated Glassware provides an added measure of safety in the event of breakage
6.7 in.
Plastic safety coating
24/40
Borosilicate Glass
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit