Learn More
DWK Life Sciences Kimble™ Kontes™ Heavy Wall Recovery Flasks with Standard Bead, standard taper Outer Joint
Designed for easy recovery of reaction products. Total contents are accessible with a spatula or policeman.
$33.10 - $111.90
Specifications
Material Glass
No. per Case 1
View More Specs
Products 10
Catalog Number Mfr. No. Capacity (Metric) Standard Taper Joint Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Standard Taper Joint Price Quantity & Availability  
K608675-0224
View Documents Promotion Details
DWK Life Sciences
6086750224
100 mL 24/25
Case for $56.80
Add to cart
K608675-0624
View Documents Promotion Details
DWK Life Sciences
6086750624
500 mL 24/25
Case for $111.90
Add to cart
K294300-0100
View Documents Promotion Details
DWK Life Sciences
2943000100
100 mL 14/20
Case for $50.80
Add to cart
K294305-0025
View Documents Promotion Details
DWK Life Sciences
2943050025
25 mL 19/22
Case for $33.10
Add to cart
K294305-0050
View Documents Promotion Details
DWK Life Sciences
2943050050
50 mL 19/22
Case for $49.00
Add to cart
K294300-0050
View Documents Promotion Details
DWK Life Sciences
2943000050
50 mL 14/20
Case for $62.40
Add to cart
K608675-0324
View Documents Promotion Details
DWK Life Sciences
6086750324
200 mL 24/25
Case for $78.60
Add to cart
K294300-0200
View Documents Promotion Details
DWK Life Sciences
2943000200
200 mL 14/20
Case for $54.85
Add to cart
K294300-0010
View Documents Promotion Details
DWK Life Sciences
2943000010
10 mL 14/20
Case for $47.45
Add to cart
K294300-0025
View Documents Promotion Details
DWK Life Sciences
2943000025
25 mL 14/20
Case for $48.05
Add to cart
Description
Description
  • Single-neck flask with standard taper outer joint
  • Designed for easy recovery of reaction products
  • Total contents are accessible with a spatula or policeman
  • Lower portion fits standard heating mantles
  • Even wall thickness and minimum runout
Specifications
Specifications
Glass
1