Learn More
DWK Life Sciences Kimble™ Kontes™ KIMAX™ Reusable Threaded Centrifuge Tubes Available on GSA/VA Contract for Federal Government customers only.
Heavy wall withstands high centrifuge speeds
$246.00 - $357.00
Specifications
Certifications/Compliance USP, ASTM E 438
Diameter (Metric) Outside 29mm
Material Borosilicate Glass
Max. Force 3,550 x g
Label Type Marking Spot
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (Metric) Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Length (Metric) Price Quantity & Availability  
05-558-12A Available on GSA/VA Contract for Federal Government customers only.
View Documents
DWK Life Sciences
4521235
35mL 100mm
Case of 12 for $246.00
Add to cart
05-558-12B Available on GSA/VA Contract for Federal Government customers only.
View Documents
DWK Life Sciences
4521250
50mL 123mm
Case of 12 for $357.00
Add to cart
Description
Description
Heavy wall withstands high centrifuge speeds
Ideal for separating neutralized sediment for culturing, from specimens of sputum in clinical work and microbiological applications.
  • Marking spot is sandblasted
  • Caps are not attached
  • Supplied with autoclavable caps with rubber liners
  • GPI thread finish is 24mm-410
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
USP, ASTM E 438
Borosilicate Glass
Marking Spot
Round
Useful in separating neutralized sediment for culturing from specimens of sputum in clinical work and microbiological applications
29mm
3,550 x g
Rubber
24-410