Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Heavy-Duty Serum Bottles
Click to view available options
Capacity (Metric):
4000 mL
9000 mL
Description
- Rounded shoulders
- Neck is tooled for a uniform fit with a No.8 rubber stopper
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Closure Material | Rubber |
Closure Size | No. 8 |
Diameter (Metric) | 167 mm |
Product Type | Serum Bottle |
Height (English) | 12.2 in. |
Height (Metric) | 310 mm |
Capacity | 135.2 oz., 4000mL |
Capacity (Metric) | 4000 mL |
Capacity (English) | 135.2 oz. |
Diameter (English) | 6.57 in. |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction