Learn More
DWK Life Sciences Kimble™ Kontes™ NMR Polyethylene Sample Tube Pressure Caps for 5 and 10mm Tubes
SDP
Tight-fitting, polyethylene cap which stays in place even with a build-up of internal vaporization pressures
$23.25 - $548.00
Specifications
Product Type Pressure Cap
Material Polyethylene
For Use With (Application) Provides tight-fitting and stays in place even with a build-up of internal vaporization pressures
View More Specs
Products 9
Catalog Number Mfr. No. Color For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. Color For Use With (Equipment) Price Quantity & Availability  
K897095-0021
SDP
View Documents Promotion Details
DWK Life Sciences
8970950021
Black 5 mm NMR Sample Tube
Case for $29.50
Add to Cart
K897095-0081
SDP
View Documents Promotion Details
DWK Life Sciences
8970950081
Yellow 5 mm NMR Sample Tube
Case for $29.50
Add to Cart
K897095-0061
SDP
View Documents Promotion Details
DWK Life Sciences
8970950061
White 5 mm NMR Sample Tube
Case for $29.50
Add to Cart
K897095-1001
SDP
View Documents Promotion Details
DWK Life Sciences
8970951001
Red 10 mm NMR Sample Tube
Case for $45.50
Add to Cart
K897095-1021
SDP
View Documents Promotion Details
DWK Life Sciences
8970951021
Black 10 mm NMR Sample Tube
Case for $45.50
Add to Cart
K8970930001
SDP
View Documents Promotion Details
DWK Life Sciences
8970930001
Red 3 mm NMR Sample Tube
Case for $23.25
Add to Cart
K897095-0001
SDP
View Documents Promotion Details
DWK Life Sciences
8970950001
Red 5 mm NMR Sample Tube
Case for $29.50
Add to Cart
K897095-0012
SDP
View Documents Promotion Details
DWK Life Sciences
8970950012
Assorted 5 mm NMR Sample Tube
Case of 600 for $548.00
Add to Cart
K897095-0071
SDP
View Documents Promotion Details
DWK Life Sciences
8970950071
Green 5 mm NMR Sample Tube
Case for $29.50
Add to Cart
Description
Description
Specifications
Specifications
Pressure Cap
Polyethylene
Provides tight-fitting and stays in place even with a build-up of internal vaporization pressures