Learn More
DWK Life Sciences Kimble™ Kontes™ Pear-Shaped Heavy Wall Boiling Flask
Single-neck flasks with a Standard Taper outer joint at the top manufactured from 33 expansion, low extractable borosilicate glass.
Supplier: DWK Life Sciences 2942500100
Description
- Available with a size 14/20, 19/22 or 24/40 taper joint
- Heavy wall flasks in a range of capacities, from 5mL to 500mL
- Conforms to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
100 mL | |
Borosilicate Glass | |
14/20 |