Kimble™ Kontes™ Polypropylene Screw Thread with Open Top
Manufacturer: Kimble™ 73806A15425
 View more versions of this product
Catalog No. 03-340-124A
  Check Availability
Add to Cart
    Specifications
    Diameter (Metric) Opening 0.87cm
    Color Black
    Material Polypropylene
    GPI Thread 15-425
    Type Open Top Screw Cap
    Material Closure Polypropylene
    Material (Liner) PTFE/Silicone
    Autoclavable Yes
    Thread Size 15-425
    Diameter (English) Opening 0.342 in.
  • Description & Specifications
Related Products
Product Content Feedback