Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Reaction-Jacketed Beakers
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Description
The open top provides easy access to concentrated residual samples.
- A conical lower interior surface eliminates spin-out when used with an egg-shaped PTFE magnetic stir bar
- Jacket has serrated 3/8 in. hose connections
- Design meets ASTM Specification E960, Type I requirements
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Specifications
Description | Connection: 0.37 in. Serrated Hose |
Diameter (English) Inner | 4.88 in. |
Diameter (English) Outer | 7 in. |
Diameter (Metric) Inner | 12.4 cm |
Diameter (Metric) Outer | 17.8 cm |
Capacity (Metric) | 2000 mL |
Material | Borosilicate Glass |
For Use With (Application) | Laboratory Applications Requiring a Variety of Temperature-Controlled Reactions and Useful in Biological and Chemical Reactions Including Titrations, Enzyme Digestions and Synthetic Organic Reactions |
Height (English) Inner | 7.48 in. |
Height (English) Outer | 8.58 in. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction