Learn More
DWK Life Sciences Kimble™ Kontes™ Reflux Condensers with Coil Type and Joints
With an internal, coil-type cold finger; Tightly wound coil
$413.00 - $472.50
Specifications
Certifications/Compliance USP Type I and ASTM E438, Type I, Class A
Material Borosilicate Glass
For Use With (Application) Condense high vapor pressure solvents like hexane
Type Reflux Condenser
Quantity Each
View More Specs
Products 2
Catalog Number Mfr. No. Height (Metric) Height (English) Price Quantity & Availability  
Catalog Number Mfr. No. Height (Metric) Height (English) Price Quantity & Availability  
K457000-0125
View Documents
DWK Life Sciences
4570000125
290 mm 11.41 in.
Case for $413.00
Add to cart
K457000-0175
View Documents
DWK Life Sciences
4570000175
340 mm 13.38 in.
Case for $472.50
Add to cart
Description
Description
  • 24/40 joints
  • 175mm cold finger coil length
  • 170cm2 condensing area
  • No. 2 hose connection
Specifications
Specifications
USP Type I and ASTM E438, Type I, Class A
Condense high vapor pressure solvents like hexane
Each
Borosilicate Glass
Reflux Condenser
24/40