Learn More
DWK Life Sciences Kimble™ Kontes™ Replacement PTFE/Silicone Septa
SDP
For Kontes brand threaded NMR tubes K897635-0800 and K897735-1300, and open-top caps.
Supplier:  DWK Life Sciences 7741610924
 View more versions of this product
Catalog No. 03-338-251
Add to Cart
Description
Description
  • Excellent for use with open-top caps to access container contents with a syringe
  • Highly chemically resistant PTFE facing maintains integrity of contents
  • Silicone rubber backing allows repeated puncturing through the seal
  • Resists coring
  • Autoclavable
  • Recommended for use in all autosamplers using screw thread vials
Specifications
Specifications
Septa
Highly chemically resistant PTFE facing maintains integrity of contents, silicone rubber backing allows repeated puncturing through the seal
Kontes™ K8976350800 and K897735130 Threaded NMR Tubes and 24 mm Threaded Open Top Cap, All autosamplers using screw thread vials
PTFE Silicone Rubber
0.010 in. (PTFE), 0.090 in. (Silicon Rubber)
Product Suggestions
Product Suggestions