Learn More
Kimble™ Kontes™ Screw Thread Culture Tubes with Unattached Rubber Lined Caps
Uniform wall thickness for maximum heat transfer and heat resistance
Manufacturer:  Kimble™ KC4506613100
 View more versions of this product
Catalog No. 12-141-356
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Phenolic
13mm
Screw
Borosilicate Glass
For biohazard test protocols
13-415
8mL
USP Type I, ASTM E 438 Type I, Class A
100mm
No
Unattached cap