Kimble™ Kontes™ Screw Thread Culture Tubes with Unattached Rubber Lined Caps

Uniform wall thickness for maximum heat transfer and heat resistance

Manufacturer: Kimble™ KC4506625150

 View more versions of this product

Catalog No. 12-141-359

  Check Availability
Add to Cart

    Specifications

    Closure Material Phenolic
    Certifications/Compliance USP Type I, ASTM E 438 Type I, Class A
    Closure Type Screw
    Capacity (Metric) 50mL
    Length (Metric) 150mm
    Material Borosilicate Glass
    Disposable No
    For Use With (Application) For biohazard test protocols
    Includes Unattached cap
    Thread Size 24-410
    Diameter (Metric) Outer 25mm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit