Kimble™ Kontes™ Serum Vials KG-33 Borosilicate Glass with Aluminum Seal

Manufacturer: Kimble™ 62121D20

 View more versions of this product

Catalog No. 10-310-242B

  Check Availability
Add to Cart

    Specifications

    Color Clear
    Material Borosilicate Glass
    Diameter (Metric) Outside 30mm
    Certifications/Compliance ASTM E438, Type I, Class A
    Type Serum Vial
    Volume (Metric) 20mL
    Height (Metric) 57mm
    Capacity (Metric) 20.0mL
    For Use With (Application) Freeze drying applications
    Classification A
    Material (Seal) Aluminum
    Volume (English) 5.4 dr.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit