Kimble™ Kontes™ Straight Connecting Adapters
Manufacturer: Kimble™ 1661003024
 View more versions of this product
Catalog No. 22-311903
  Check Availability
Add to Cart
    Specifications
    Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
    Length (English) 4.33 in.
    Material Borosilicate Glass
    Product Type Straight Connecting Adapter
    Standard Taper Joint No. 24/40
    For Use With (Equipment) Useful as an air condenser
    Length (Metric) 110mm
  • Description & Specifications
Related Products
Product Content Feedback