Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KOR Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11079876H
Description
KOR Polyclonal antibody specifically detects KOR in Human, Rat samples. It is validated for Western BlotSpecifications
KOR | |
Polyclonal | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Western Blot | |
HRP | |
kappa opioid receptor, kappa-type opioid receptor, KOR, K-OR-1, KOR-1, Opiate receptor, kappa-1, opioid receptor, kappa 1, OPRK | |
Synthetic peptide comprising residues 366-380 of the human KOR-1 protein. Sequence identity is 66% in mouse and rat. | |
0.1 mL | |
GPCR, Signal Transduction | |
4986 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction