Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kos1/Tnk1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$177.16 - $378.00
Specifications
Antigen | Kos1/Tnk1 |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Description
Kos1/Tnk1 Polyclonal specifically detects Kos1/Tnk1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Kos1/Tnk1 | |
Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Polyclonal | |
Rabbit | |
Human, Mouse | |
Q99ML2 | |
IgG | |
Affinity Purified | |
This is specific for Kos1/Tnk1 |
1 mg/mL | |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
PBS with 0.02% Sodium Azide | |
C-terminal residues 421-434 [QNQVQSLPAGSAEP] of the 47 kDa mouse Kos1 Tnk1 protein. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title