Learn More
Leap PAL PartsSupplier Diversity Partner Center Draining, Screw Vial, Clear, Kit Pack, 100 Pack  DFS Item
Manufacturer:  Leap PAL PartsSupplier Diversity Partner XQS2009
Kit Pack. CD Vial, 1.5-mL, 12x32mm, Screw, Clear, 9mm, Blue QuikCap, pre-slit, PTFE/Silicone, 100 Pack
Catalog No. 50-635-212