Learn More
Leap PAL PartsSupplier Diversity Partner Clear Screw Kit Pack 2mL 12x32mm Pico Pure Cap, 100 Pack  DFS Item
Manufacturer:  Leap PAL PartsSupplier Diversity Partner XAR6088
Kit Pack, Screw, Clear, 12x32mm, Flat bottom, Pico Pure Cap, starburst cut, 100 Pack
Catalog No. 50-635-189