Learn More
Leap PAL PartsSupplier Diversity Partner Kit Pack Snap 700l 12x32mm Flat, 100 Pack  DFS Item
Manufacturer:  Leap PAL PartsSupplier Diversity Partner XAR6066
Kit Pack, Snap, LC/GC, With Marking Panel (No), 12x32mm, Flat bottom, Quantity100 Pack No, QuikCap/QuiKVial
Catalog No. 50-635-207