Learn More
Leap PAL Parts KitPack, Screw, Amber, 4mL, 15x45, Pre-slit PTFE/SIL, 100 Pack
Supplier:  Leap PAL Parts XAR1674
Encompass_Preferred
Kit Pack. 4-mL, 15x45mm, Screw, Amber, pre-slit, PTFE/Silicone, Bonded, 40-mil, 100 Pack
Catalog No. 50-635-202