Learn More
Leap PAL PartsSupplier Diversity Partner Kit Pack Screw Amber 1.0mL 13x41mm Flat, 12 Pack  DFS Item
Manufacturer:  Leap PAL PartsSupplier Diversity Partner XQS2302
Kit Pack. CD Vial, Heavy-walled, 1.0-mL, 13x41mm, Screw, Amber, 13-425, PTFE/Silicone, 12 Pack
Catalog No. 50-635-244