Learn More
Leap PAL PartsSupplier Diversity Partner Kit Pack Screw Clear 2.0mL 20x40mm Flat, 12 Pack  DFS Item
Manufacturer:  Leap PAL PartsSupplier Diversity Partner XQS2266
Kit Pack. CD Vial, Heavy-walled, Graduated, 2.0-mL, 20x40mm, Screw, Clear, 20-400, PTFE/Silicone, 12 Pack
Catalog No. 50-635-241